Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Ethylhexyl bromide, 96%, stab. with copper

Click to view available options
Quantity:
25g
100g
500g
18908-66-2
C8H17Br
193.128
MFCD00000220
NZWIYPLSXWYKLH-UHFFFAOYSA-N
2-ethylhexyl bromide,3-bromomethyl heptane,heptane, 3-bromomethyl,1-bromo-2-ethylhexane,1-bromo iso octane,2-ethyl-bromohexane,3-bromomethyl-heptane,1-bromo-2 ethylhexane,2-ethylhexylhydrobromide
86804
3-(bromomethyl)heptane
CCCCC(CC)CBr
18908-66-2
C8H17Br
193.128
MFCD00000220
NZWIYPLSXWYKLH-UHFFFAOYSA-N
2-ethylhexyl bromide,3-bromomethyl heptane,heptane, 3-bromomethyl,1-bromo-2-ethylhexane,1-bromo iso octane,2-ethyl-bromohexane,3-bromomethyl-heptane,1-bromo-2 ethylhexane,2-ethylhexylhydrobromide
86804
3-(bromomethyl)heptane
CCCCC(CC)CBr
Applications
2-Ethylhexyl bromide is used in the synthesis of methoxy-4-(2-ethylhexyloxy)benzene, 1-bromo-4-(2′-ethylhexyloxy)benzene and 3,6-diaryl substituted 9-alkylcarbazoles as hole transporting materials for various organic light emitting devices.

Solubility
Soluble in chloroform, ethyl acetate. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from heat, oxidizing agents.
Boiling Point 94°C to 95°C (35mmHg)
Flash Point 69°C (156°F)
Quantity 25g
Beilstein 1098313
Formula Weight 193.13
Percent Purity 96%
Chemical Name or Material 2-Ethylhexyl bromide, stabilized with copper

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Recommended Storage : 2° to 8°C