Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-Ethylhexylamine, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Description
2-Ethylhexylamine is used to make pharmaceuticals and other chemicals. It is used as beam material and surface active agent. It also involves in synthesis of aliphatic amines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Ethylhexylamine is used to make pharmaceuticals and other chemicals. It is used as beam material and surface active agent. It also involves in synthesis of aliphatic amines.
Solubility
Soluble in water. 2.5 g/L (20°C)
Notes
Air Sensitive. Incompatible with strong oxidizing agents. Protect against electrostatic charges. Store away from acid chlorides and acid anhydrides.
2-Ethylhexylamine is used to make pharmaceuticals and other chemicals. It is used as beam material and surface active agent. It also involves in synthesis of aliphatic amines.
Solubility
Soluble in water. 2.5 g/L (20°C)
Notes
Air Sensitive. Incompatible with strong oxidizing agents. Protect against electrostatic charges. Store away from acid chlorides and acid anhydrides.
Specifications
Melting Point | -76°C |
Density | 0.789 |
Boiling Point | 169°C |
Flash Point | 50°C (122°F) |
Odor | Amine-like |
Refractive Index | 1.43 |
Quantity | 500 mL |
UN Number | UN2276 |
Beilstein | 1209249 |
Sensitivity | Air Sensitive |
Show More |
Safety and Handling
Hazard Category | H226-H302-H312-H314-H318-H331 |
Hazard Statement | GHS H Statement H311-H314-H318-H226-H302 Toxic in contact with skin. Causes severe skin burns and eye damage. Causes serious eye damage. Flammable liquid and vapor. Harmful if swallowed. |
Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P312-P302+P352-P303+P361+P353-P304+P340-P305+P351+P338-P310-P311-P330-P362-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: 2-ETHYLHEXYLAMINE |
EINECSNumber | 203-233-8 |
RTECSNumber | MQ5250000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction