Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethylpyridine, 98+%, ACROS Organics™

Click to view available options
Quantity:
500g
100-71-0
C7H9N
107.156
MFCD00006361
NRGGMCIBEHEAIL-UHFFFAOYSA-N
pyridine, 2-ethyl,ethylpyridine,2-ethyl-pyridine,pyridine, ethyl,alpha-ethylpyridine,.alpha.-ethylpyridine,unii-06x1w46pyx,2-ethylpyridin,2-ethylpyridine,acmc-2097ry
7523
2-ethylpyridine
CCC1=CC=CC=N1
100-71-0
C7H9N
107.156
MFCD00006361
NRGGMCIBEHEAIL-UHFFFAOYSA-N
pyridine, 2-ethyl,ethylpyridine,2-ethyl-pyridine,pyridine, ethyl,alpha-ethylpyridine,.alpha.-ethylpyridine,unii-06x1w46pyx,2-ethylpyridin,2-ethylpyridine,acmc-2097ry
7523
2-ethylpyridine
CCC1=CC=CC=N1
Density 0.937g/mL
Infrared Spectrum Authentic
Refractive Index 1.4960 to 1.4980 (20°C, 589nm)
Quantity 500g
Color Colorless to Yellow
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Linear Formula (C5H4N)CH2CH3
Beilstein 20, 241
Formula Weight 107.15
Physical Form Liquid
Percent Purity ≥98.0%
Chemical Name or Material 2-Ethylpyridine, 98+%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 202-881-9

TSCA : TSCA