Alfa Aesar™ 2-(Ethylthio)ethylamine, 97%

Manufacturer: Alfa Aesar™ B2214706

Quantity 5g

 View more versions of this product

Catalog No. AAB2214706

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 36489-03-9
    Molecular Formula CH3CH2SCH2CH2NH2
    Formula Weight 105.2
    MDL Number MFCD00014826
    UN Number UN2733
    Beilstein 1731375
    Chemical Name or Material 2-(Ethylthio)ethylamine

    Specifications

    Formula Weight 105.2
    MDL Number MFCD00014826
    Health Hazard 1 Danger
    Flash Point 52°C (125°F)
    Boiling Point 159° to 160°C
    Quantity 5g
    Molecular Formula CH3CH2SCH2CH2NH2
    CAS 36489-03-9
    UN Number UN2733
    Beilstein 1731375
    Health Hazard 2 GHS H Statement
    H314-H318-H226
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable liquid and vapour.
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
    Chemical Name or Material 2-(Ethylthio)ethylamine
    Assay 97%

    GHS H Statement
    H314-H318-H226
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable liquid and vapour.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit