Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Fluoro-3-hydroxy-6-nitrophenylacetone, 97%

Click to view available options
Quantity:
250mg
1g
649736-31-2
C9H8FNO4
213.164
MFCD11869874
IPRZVDNMKMONNN-UHFFFAOYSA-N
1-2-fluoro-3-hydroxy-6-nitrophenyl propan-2-one,2-propanone, 1-2-fluoro-3-hydroxy-6-nitrophenyl,1-2-fluoro-3-hydroxy-6-nitrophenyl propan-1-one,3-acetylmethyl-2-fluoro-4-nitrophenol,1-2-fluoro-3-hydroxy-6-nitrophenyl-propan-2-one,1-2-fluoro-3-hydroxy-6nitrophenyl-propan-2-one,1-2-fluoro-3-hydroxy-6-nitro-phenyl-propan-1-one
22337995
CC(=O)CC1=C(C=CC(=C1F)O)[N+](=O)[O-]
649736-31-2
C9H8FNO4
213.164
MFCD11869874
IPRZVDNMKMONNN-UHFFFAOYSA-N
1-2-fluoro-3-hydroxy-6-nitrophenyl propan-2-one,2-propanone, 1-2-fluoro-3-hydroxy-6-nitrophenyl,1-2-fluoro-3-hydroxy-6-nitrophenyl propan-1-one,3-acetylmethyl-2-fluoro-4-nitrophenol,1-2-fluoro-3-hydroxy-6-nitrophenyl-propan-2-one,1-2-fluoro-3-hydroxy-6nitrophenyl-propan-2-one,1-2-fluoro-3-hydroxy-6-nitro-phenyl-propan-1-one
22337995
CC(=O)CC1=C(C=CC(=C1F)O)[N+](=O)[O-]
Quantity 250mg
Beilstein 11804446
Formula Weight 213.16
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 2-Fluoro-3-hydroxy-6-nitrophenylacetone

storageNote1 : 2° to 8°C