Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoro-3-iodopyridine, 95%, Acros Organics™

Click to view available options
Quantity:
1g
113975-22-7
C5H3FIN
222.98
WCDCAXVNBOLWNO-UHFFFAOYSA-N
2-fluoro-3-iodo-pyridine,pyridine, 2-fluoro-3-iodo,zlchem 409,2-fluoro-3 iodopyridine,2-fluoro-3-iodo-pyridin,2-fluoro-3-iodopyridine,2-fluoro-3-iodopyridine;,2-fluoro-3-iodo pyridine,acmc-1c73w,ksc174s0n
7009501
2-fluoro-3-iodopyridine
C1=CC(=C(N=C1)F)I
113975-22-7
C5H3FIN
222.98
WCDCAXVNBOLWNO-UHFFFAOYSA-N
2-fluoro-3-iodo-pyridine,pyridine, 2-fluoro-3-iodo,zlchem 409,2-fluoro-3 iodopyridine,2-fluoro-3-iodo-pyridin,2-fluoro-3-iodopyridine,2-fluoro-3-iodopyridine;,2-fluoro-3-iodo pyridine,acmc-1c73w,ksc174s0n
7009501
2-fluoro-3-iodopyridine
C1=CC(=C(N=C1)F)I
Packaging Glass Bottle
Quantity 1g
Melting Point 45.0°C to 47.5°C
Assay Percent Range 95%
Formula Weight 222.98
Percent Purity 95%
Chemical Name or Material 2-Fluoro-3-iodopyridine

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Warning