Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Fluoro-3-nitrobenzotrifluoride, 98%

Click to view available options
Quantity:
250mg
1g
5g
61324-97-8
C7H3F4NO2
209.1
MFCD08532487
PGKWPKDZNHZBQK-UHFFFAOYSA-N
2-fluoro-3-nitrobenzotrifluoride,2-fluoro-1-nitro-3-trifluoromethyl benzene,benzene, 2-fluoro-1-nitro-3-trifluoromethyl,2-fluoro-3-trifluoromethyl nitrobenzene,2-fluoro-3-trifluoromethylnitrobenzene,2-fluoro-3-nitrobenzotrifluori
23340645
2-fluoro-1-nitro-3-(trifluoromethyl)benzene
C1=CC(=C(C(=C1)[N+](=O)[O-])F)C(F)(F)F
61324-97-8
C7H3F4NO2
209.1
MFCD08532487
PGKWPKDZNHZBQK-UHFFFAOYSA-N
2-fluoro-3-nitrobenzotrifluoride,2-fluoro-1-nitro-3-trifluoromethyl benzene,benzene, 2-fluoro-1-nitro-3-trifluoromethyl,2-fluoro-3-trifluoromethyl nitrobenzene,2-fluoro-3-trifluoromethylnitrobenzene,2-fluoro-3-nitrobenzotrifluori
23340645
2-fluoro-1-nitro-3-(trifluoromethyl)benzene
C1=CC(=C(C(=C1)[N+](=O)[O-])F)C(F)(F)F
Quantity 250mg
UN Number UN2811
Beilstein 13320950
Formula Weight 209.1
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 2-Fluoro-3-nitrobenzotrifluoride

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.