Alfa Aesar™ 2-Fluoro-3-(trifluoromethyl)benzaldehyde, 97%

Manufacturer: Alfa Aesar™ B2373803

Quantity 1g

 View more versions of this product

Catalog No. AAB2373803

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 112641-20-0
    Molecular Formula C8H4F4O
    Formula Weight 192.11
    MDL Number MFCD00061246
    Beilstein 8311752
    Synonym alpha,alpha,alpha,2-Tetrafluoro-m-tolualdehyde
    Chemical Name or Material 2-Fluoro-3-(trifluoromethyl)benzaldehyde

    Specifications

    Formula Weight 192.11
    Molecular Formula C8H4F4O
    MDL Number MFCD00061246
    CAS 112641-20-0
    Beilstein 8311752
    Synonym alpha,alpha,alpha,2-Tetrafluoro-m-tolualdehyde
    Health Hazard 1 Warning
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Flash Point 73°C (163°F)
    Quantity 1g
    Health Hazard 2 GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.
    Chemical Name or Material 2-Fluoro-3-(trifluoromethyl)benzaldehyde
    Assay 97%

    GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.

    GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit