Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Fluoro-3-(trifluoromethyl)benzaldehyde, 97%

Click to view available options
Quantity:
1g
5g
112641-20-0
C8H4F4O
192.113
MFCD00061246
XDMZVNQKVMTCSP-UHFFFAOYSA-N
2-fluoro-3-trifluoromethyl benzaldehyde,benzaldehyde, 2-fluoro-3-trifluoromethyl,2-fluoro-3-trifluoromethyl-benzaldehyde,pubchem7149,acmc-1c9j0,attercop-chm at111493,a, a, a,2-tetrafluoro-m-tolualdehyde,benzaldehyde,2-fluoro-3-trifluoromethyl
517978
2-fluoro-3-(trifluoromethyl)benzaldehyde
C1=CC(=C(C(=C1)C(F)(F)F)F)C=O
112641-20-0
C8H4F4O
192.113
MFCD00061246
XDMZVNQKVMTCSP-UHFFFAOYSA-N
2-fluoro-3-trifluoromethyl benzaldehyde,benzaldehyde, 2-fluoro-3-trifluoromethyl,2-fluoro-3-trifluoromethyl-benzaldehyde,pubchem7149,acmc-1c9j0,attercop-chm at111493,a, a, a,2-tetrafluoro-m-tolualdehyde,benzaldehyde,2-fluoro-3-trifluoromethyl
517978
2-fluoro-3-(trifluoromethyl)benzaldehyde
C1=CC(=C(C(=C1)C(F)(F)F)F)C=O
Quantity 5g
Flash Point 73°C (163°F)
Beilstein 8311752
Formula Weight 192.11
Percent Purity 97%
Chemical Name or Material 2-Fluoro-3-(trifluoromethyl)benzaldehyde

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning