Alfa Aesar™ 2-Fluoro-3-(trifluoromethyl)benzoyl chloride, 97%

Manufacturer: Alfa Aesar™ B2346406

Quantity 5g

 View more versions of this product

Catalog No. AAB2346406

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 208173-19-7
    Molecular Formula C8H3ClF4O
    Formula Weight 226.56
    MDL Number MFCD00061154
    UN Number UN3265
    Beilstein 7992377
    Synonym alpha,alpha,alpha,2-Tetrafluoro-m-toluoyl chloride
    Chemical Name or Material 2-Fluoro-3-(trifluoromethyl)benzoyl chloride

    Specifications

    Formula Weight 226.56
    Molecular Formula C8H3ClF4O
    UN Number UN3265
    Synonym alpha,alpha,alpha,2-Tetrafluoro-m-toluoyl chloride
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
    Flash Point 101°C (213°F)
    Boiling Point 192° to 194°C
    MDL Number MFCD00061154
    CAS 208173-19-7
    Beilstein 7992377
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P309-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Quantity 5g
    Chemical Name or Material 2-Fluoro-3-(trifluoromethyl)benzoyl chloride
    Assay 97%

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P280-P305+P351+P338-P309-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit