Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Fluoro-4-(4-methyl-1-piperazinylcarbonyl)benzeneboronic acid, 96%

Click to view available options
Quantity:
250mg
1g
1454901-23-5
1454901-23-5
Quantity 1g
Color Brown
Beilstein 24007287
Formula Weight 266.08
Physical Form Crystals
Percent Purity 96%
Chemical Name or Material 2-Fluoro-4-(4-methyl-1-piperazinylcarbonyl)benzeneboronic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning