Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Fluoro-4,6-bis(trifluoromethyl)benzoyl chloride, 97%

Manufacturer:  Alfa Aesar™ H3229203

Quantity 1g

 View more versions of this product

Catalog No. AAH3229203


  Check Availability
Add to cart

Specifications

Description

Applications
2-Fluoro-4,6-bis(trifluoromethyl)benzoyl chloride, is used as a pharmaceutical intermediate.

Solubility
It reacts with water.

Notes
Moisture&water Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, bases.
Description & Specifications

Chemical Identifiers

C9H2ClF7O
MFCD09258670
2-Fluoro-4,6-bis(trifluoromethyl)benzoyl chloride
294.55
UN3265

Specifications

1g
294.55
UN3265
97%
C9H2ClF7O
MFCD09258670
2-Fluoro-4,6-bis(trifluoromethyl)benzoyl chloride

Safety and Handling

SDS & Safety Documents

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit