Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-Fluoro-4-hydroxybenzoic acid, 98%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
25 g
Specifications
Melting Point | 187.0°C to 188.0°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (HPLC) |
Quantity | 25 g |
Formula Weight | 156.11 |
Percent Purity | 98% |
Chemical Name or Material | 2-Fluoro-4-hydroxybenzoic acid |
Safety and Handling
Hazard Category | GHS Signal Word: Warning |
Hazard Statement | GHS H Statement Causes skin irritation. Causes serious eye irritation. |
Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse. |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction