Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Fluoro-4-hydroxybenzonitrile, 99%

Click to view available options
Quantity:
1g
5g
25g
82380-18-5
C7H4FNO
137.113
MFCD00192177
REIVHYDACHXPNH-UHFFFAOYSA-N
4-cyano-3-fluorophenol,3-fluoro-4-cyanophenol,2-fluoro-4-benzonitrile,benzonitrile, 2-fluoro-4-hydroxy,4-hydroxy-2-fluorobenzonitrile,2-fluoro-4-hydroxy-benzonitrile,2-fluoro-4-hydroxybenzenecarbonitrile,pubchem2610,pubchem3495
2734675
2-fluoro-4-hydroxybenzonitrile
C1=CC(=C(C=C1O)F)C#N
82380-18-5
C7H4FNO
137.113
MFCD00192177
REIVHYDACHXPNH-UHFFFAOYSA-N
4-cyano-3-fluorophenol,3-fluoro-4-cyanophenol,2-fluoro-4-benzonitrile,benzonitrile, 2-fluoro-4-hydroxy,4-hydroxy-2-fluorobenzonitrile,2-fluoro-4-hydroxy-benzonitrile,2-fluoro-4-hydroxybenzenecarbonitrile,pubchem2610,pubchem3495
2734675
2-fluoro-4-hydroxybenzonitrile
C1=CC(=C(C=C1O)F)C#N
Density 1.45
Melting Point 123°C to 126°C
Quantity 25g
UN Number UN3077
Beilstein 4741066
Formula Weight 137.11
Percent Purity 99%
Chemical Name or Material 2-Fluoro-4-hydroxybenzonitrile

GHS H Statement
H301-H318
Toxic if swallowed.
Causes serious eye damage.

P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c

H302-H318

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

EINECSNumber : 422-810-7

TSCA : No

Recommended Storage : Ambient temperatures