Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoro-4-hydroxybenzonitrile, 99%, Acros Organics

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
82380-18-5
C7H4FNO
137.113
MFCD00192177
REIVHYDACHXPNH-UHFFFAOYSA-N
4-cyano-3-fluorophenol,3-fluoro-4-cyanophenol,2-fluoro-4-benzonitrile,benzonitrile, 2-fluoro-4-hydroxy,4-hydroxy-2-fluorobenzonitrile,2-fluoro-4-hydroxy-benzonitrile,2-fluoro-4-hydroxybenzenecarbonitrile,pubchem2610,pubchem3495
2734675
2-fluoro-4-hydroxybenzonitrile
C1=CC(=C(C=C1O)F)C#N
82380-18-5
C7H4FNO
137.113
MFCD00192177
REIVHYDACHXPNH-UHFFFAOYSA-N
4-cyano-3-fluorophenol,3-fluoro-4-cyanophenol,2-fluoro-4-benzonitrile,benzonitrile, 2-fluoro-4-hydroxy,4-hydroxy-2-fluorobenzonitrile,2-fluoro-4-hydroxy-benzonitrile,2-fluoro-4-hydroxybenzenecarbonitrile,pubchem2610,pubchem3495
2734675
2-fluoro-4-hydroxybenzonitrile
C1=CC(=C(C=C1O)F)C#N
Quantity 5g
Infrared Spectrum Authentic
Packaging Glass bottle
Color White
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Formula Weight 137.11
Physical Form Fine Crystalline Needles or Powder
Percent Purity ≥98.5%
Chemical Name or Material 2-Fluoro-4-hydroxybenzonitrile, 99%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Toxic to aquatic life with long lasting effects.
Causes serious eye damage.

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger