Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Fluoro-4-iodo-5-methylpyridine, 95%

Click to view available options
Quantity:
250mg
1g
5g
153034-94-7
C6H5FIN
237.016
MFCD03095297
BVKQLNXPPQEELX-UHFFFAOYSA-N
2-fluoro-4-iodo-5-picoline,2-fluoro-4-iodo-5-methyl-pyridine,6-fluoro-4-iodo-b-picoline,6-fluoro-4-iodo-3-picoline,2-fluoro-4-iodo-5-methyl pyridine,pyridine,2-fluoro-4-iodo-5-methyl,pyridine, 2-fluoro-4-iodo-5-methyl,pubchem1305,acmc-20el7f,abbypharma ap-30-1289
10060052
2-fluoro-4-iodo-5-methylpyridine
CC1=CN=C(C=C1I)F
153034-94-7
C6H5FIN
237.016
MFCD03095297
BVKQLNXPPQEELX-UHFFFAOYSA-N
2-fluoro-4-iodo-5-picoline,2-fluoro-4-iodo-5-methyl-pyridine,6-fluoro-4-iodo-b-picoline,6-fluoro-4-iodo-3-picoline,2-fluoro-4-iodo-5-methyl pyridine,pyridine,2-fluoro-4-iodo-5-methyl,pyridine, 2-fluoro-4-iodo-5-methyl,pubchem1305,acmc-20el7f,abbypharma ap-30-1289
10060052
2-fluoro-4-iodo-5-methylpyridine
CC1=CN=C(C=C1I)F
Quantity 5g
Color White
Sensitivity Light Sensitive
Melting Point 40°C to 45°C
Beilstein 6643371
Formula Weight 237.02
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 2-Fluoro-4-iodo-5-methylpyridine

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning