Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Fluoro-4-iodoaniline, 99%

Click to view available options
Quantity:
5g
25g
100g
29632-74-4
C6H5FIN
237.016
MFCD00011738
CUMTUBVTKOYYOU-UHFFFAOYSA-N
2-fluoro-4-iodo-phenylamine,benzenamine, 2-fluoro-4-iodo,2-fluoro-4-iodobenzenamine,2-fluoro-4-iodo aniline,2-fluoro-4-iodo-aniline,2-fluoro-4-iodophenylamine,zlchem 543,pubchem1532,2-fluoro4-iodoaniline,2-fluoro-4-iodaniline
185694
2-fluoro-4-iodoaniline
C1=CC(=C(C=C1I)F)N
29632-74-4
C6H5FIN
237.016
MFCD00011738
CUMTUBVTKOYYOU-UHFFFAOYSA-N
2-fluoro-4-iodo-phenylamine,benzenamine, 2-fluoro-4-iodo,2-fluoro-4-iodobenzenamine,2-fluoro-4-iodo aniline,2-fluoro-4-iodo-aniline,2-fluoro-4-iodophenylamine,zlchem 543,pubchem1532,2-fluoro4-iodoaniline,2-fluoro-4-iodaniline
185694
2-fluoro-4-iodoaniline
C1=CC(=C(C=C1I)F)N
Applications
2-Fluoro-4-iodoaniline is widely used in the pharmaceutical, chemical and food industries as a fundamental building block to create a variety of important compounds.

Solubility
Insoluble in water.

Notes
Light sensitive. Incompatible with oxidizing agents.
Quantity 5g
Melting Point 55°C to 57°C
Flash Point >110°C (230°F)
Beilstein 2081100
Formula Weight 237.02
Percent Purity 99%
Chemical Name or Material 2-Fluoro-4-iodoaniline

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger