Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Fluoro-4-iodoaniline, 99%

Click to view available options
Quantity:
5g
25g
100g
29632-74-4
C6H5FIN
237.016
MFCD00011738
CUMTUBVTKOYYOU-UHFFFAOYSA-N
2-fluoro-4-iodo-phenylamine,benzenamine, 2-fluoro-4-iodo,2-fluoro-4-iodobenzenamine,2-fluoro-4-iodo aniline,2-fluoro-4-iodo-aniline,2-fluoro-4-iodophenylamine,zlchem 543,pubchem1532,2-fluoro4-iodoaniline,2-fluoro-4-iodaniline
185694
2-fluoro-4-iodoaniline
C1=CC(=C(C=C1I)F)N
29632-74-4
C6H5FIN
237.016
MFCD00011738
CUMTUBVTKOYYOU-UHFFFAOYSA-N
2-fluoro-4-iodo-phenylamine,benzenamine, 2-fluoro-4-iodo,2-fluoro-4-iodobenzenamine,2-fluoro-4-iodo aniline,2-fluoro-4-iodo-aniline,2-fluoro-4-iodophenylamine,zlchem 543,pubchem1532,2-fluoro4-iodoaniline,2-fluoro-4-iodaniline
185694
2-fluoro-4-iodoaniline
C1=CC(=C(C=C1I)F)N
Applications
2-Fluoro-4-iodoaniline is widely used in the pharmaceutical, chemical and food industries as a fundamental building block to create a variety of important compounds.

Solubility
Insoluble in water.

Notes
Light sensitive. Incompatible with oxidizing agents.
Sensitivity Light sensitive
Melting Point 55°C to 57°C
Quantity 25g
Flash Point >110°C (230°F)
Beilstein 2081100
Solubility Information Insoluble in water.
Formula Weight 237.02
Percent Purity 99%
Chemical Name or Material 2-Fluoro-4-iodoaniline

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

TSCA : No

Recommended Storage : Ambient temperatures