Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2'-Fluoro-4'-methoxyacetophenone, 99%

Click to view available options
Quantity:
5g
25g
100g
74457-86-6
C9H9FO2
168.167
MFCD00042290
PIRRWUMTIBFCCW-UHFFFAOYSA-N
2'-fluoro-4'-methoxyacetophenone,2-fluoro-4-methoxyacetophenone,1-2-fluoro-4-methoxyphenyl ethanone,1-2-fluoro-4-methoxyphenyl ethan-1-one,4-acetyl-3-fluoroanisole,1-acetyl-2-fluoro-4-methoxybenzene,2'-fluor-4'-methoxyacetophenone,2-fluoro-4-methoxy-acetophenone,ethanone, 1-2-fluoro-4-methoxyphenyl,zlchem 344
592821
1-(2-fluoro-4-methoxyphenyl)ethanone
CC(=O)C1=C(C=C(C=C1)OC)F
74457-86-6
C9H9FO2
168.167
MFCD00042290
PIRRWUMTIBFCCW-UHFFFAOYSA-N
2'-fluoro-4'-methoxyacetophenone,2-fluoro-4-methoxyacetophenone,1-2-fluoro-4-methoxyphenyl ethanone,1-2-fluoro-4-methoxyphenyl ethan-1-one,4-acetyl-3-fluoroanisole,1-acetyl-2-fluoro-4-methoxybenzene,2'-fluor-4'-methoxyacetophenone,2-fluoro-4-methoxy-acetophenone,ethanone, 1-2-fluoro-4-methoxyphenyl,zlchem 344
592821
1-(2-fluoro-4-methoxyphenyl)ethanone
CC(=O)C1=C(C=C(C=C1)OC)F
Quantity 100g
Melting Point 50°C to 53°C
Boiling Point 80°C to 85°C (0.1mmHg)
Flash Point >110°C (230°F)
Beilstein 3539992
Formula Weight 168.17
Percent Purity 99%
Chemical Name or Material 2'-Fluoro-4'-methoxyacetophenone

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures