Alfa Aesar™ 2-Fluoro-4-methoxybenzonitrile, 97%

Manufacturer: Alfa Aesar™ H3253103

Quantity 1g

 View more versions of this product

Catalog No. AAH3253103

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 94610-82-9
    Molecular Formula C8H6FNO
    Formula Weight 151.14
    MDL Number MFCD00509374
    UN Number UN3439
    Chemical Name or Material 2-Fluoro-4-methoxybenzonitrile

    Specifications

    Formula Weight 151.14
    Molecular Formula C8H6FNO
    MDL Number MFCD00509374
    UN Number UN3439
    Health Hazard 1 Danger
    Quantity 1g
    CAS 94610-82-9
    Health Hazard 2 GHS H Statement
    H331-H302-H312-H315-H319-H335
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
    Melting Point 62° to 65°C
    Chemical Name or Material 2-Fluoro-4-methoxybenzonitrile
    Assay 97%

    Description

    Applications
    2-Fluoro-4-methoxybenzonitrile is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

    Notes
    Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

    GHS H Statement
    H331-H302-H312-H315-H319-H335
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit