Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Fluoro-4-methyl-3-nitropyridine, 98%

Click to view available options
Quantity:
250mg
1g
5g
19346-43-1
C6H5FN2O2
156.116
MFCD04112526
AUHFWLUBEJKXLN-UHFFFAOYSA-N
2-fluoro-3-nitro-4-picoline,pyridine, 2-fluoro-4-methyl-3-nitro,2-fluoro-3-nitro-4-methylpyridine,pubchem6297,ksc541e5r,2-fluor-3-nitro-4-methylpyridin,2-fluoro-4-methyl-3-nitro-pyridine,pyridine,2-fluoro-4-methyl-3-nitro,2-fluoranyl-4-methyl-3-nitro-pyridine,2-fluoro-3-nitro-4-picoline
12408182
2-fluoro-4-methyl-3-nitropyridine
CC1=C(C(=NC=C1)F)[N+](=O)[O-]
19346-43-1
C6H5FN2O2
156.116
MFCD04112526
AUHFWLUBEJKXLN-UHFFFAOYSA-N
2-fluoro-3-nitro-4-picoline,pyridine, 2-fluoro-4-methyl-3-nitro,2-fluoro-3-nitro-4-methylpyridine,pubchem6297,ksc541e5r,2-fluor-3-nitro-4-methylpyridin,2-fluoro-4-methyl-3-nitro-pyridine,pyridine,2-fluoro-4-methyl-3-nitro,2-fluoranyl-4-methyl-3-nitro-pyridine,2-fluoro-3-nitro-4-picoline
12408182
2-fluoro-4-methyl-3-nitropyridine
CC1=C(C(=NC=C1)F)[N+](=O)[O-]
Quantity 250mg
Melting Point 33°C
Beilstein 1639010
Formula Weight 156.12
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 2-Fluoro-4-methyl-3-nitropyridine