Alfa Aesar™ 2-Fluoro-4-methylbenzotrifluoride, 97%

Manufacturer: Alfa Aesar™ H3239603

Quantity 1g

 View more versions of this product

Catalog No. AAH3239603

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 158364-83-1
    Molecular Formula C8H6F4
    Formula Weight 178.13
    MDL Number MFCD07368763
    UN Number UN1993
    Synonym 3-Fluoro-4-(trifluoromethyl)toluene
    Chemical Name or Material 2-Fluoro-4-methylbenzotrifluoride

    Specifications

    Formula Weight 178.13
    Molecular Formula C8H6F4
    MDL Number MFCD07368763
    UN Number UN1993
    Synonym 3-Fluoro-4-(trifluoromethyl)toluene
    Health Hazard 2 GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P260-P280g-P305+P351+P338
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Flash Point 39°C (102°F)
    Boiling Point 146°C
    Quantity 1g
    CAS 158364-83-1
    Health Hazard 1 Warning
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    Chemical Name or Material 2-Fluoro-4-methylbenzotrifluoride
    Assay 97%

    GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P260-P280g-P305+P351+P338
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit