Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Fluoro-4-methylbenzotrifluoride, 97%

Manufacturer:  Alfa Aesar™ H3239614

Quantity 25g

 View more versions of this product

Catalog No. AAH3239614


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

158364-83-1
178.13
UN1993
2-Fluoro-4-methylbenzotrifluoride
C8H6F4
MFCD07368763
3-Fluoro-4-(trifluoromethyl)toluene

Specifications

39°C (102°F)
25g
C8H6F4
MFCD07368763
3-Fluoro-4-(trifluoromethyl)toluene
97%
146°C
158364-83-1
178.13
UN1993
2-Fluoro-4-methylbenzotrifluoride

Safety and Handling

SDS & Safety Documents

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P260-P280g-P305+P351+P338
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit