Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoro-4-nitrotoluene, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
1427-07-2
C7H6FNO2
155.13
MFCD00007199
WIQISTBTOQNVCE-UHFFFAOYSA-N
2-fluoro-4-nitrotoluene,toluene, 2-fluoro-4-nitro,benzene, 2-fluoro-1-methyl-4-nitro,3-fluoro-4-methylnitrobenzene,1-methyl-2-fluoro-4-nitrobenzene,2-fluoro-1-methyl-4-nitro-benzene,pubchem1592,2-fluoro4-nitrotoluene,acmc-209cov,2-fluor-4-nitro-toluol
74025
2-fluoro-1-methyl-4-nitrobenzene
CC1=C(C=C(C=C1)[N+](=O)[O-])F
1427-07-2
C7H6FNO2
155.13
MFCD00007199
WIQISTBTOQNVCE-UHFFFAOYSA-N
2-fluoro-4-nitrotoluene,toluene, 2-fluoro-4-nitro,benzene, 2-fluoro-1-methyl-4-nitro,3-fluoro-4-methylnitrobenzene,1-methyl-2-fluoro-4-nitrobenzene,2-fluoro-1-methyl-4-nitro-benzene,pubchem1592,2-fluoro4-nitrotoluene,acmc-209cov,2-fluor-4-nitro-toluol
74025
2-fluoro-1-methyl-4-nitrobenzene
CC1=C(C=C(C=C1)[N+](=O)[O-])F
Quantity 5g
Color Yellow to Brown
Boiling Point 65.0°C to 68.0°C (2.0mmHg)
Flash Point 73°C
Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 33.0°C to 37.0°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula CH3C6H3(NO2)F
Beilstein 05, III, 743
Formula Weight 155.13
Physical Form Crystalline Low Melting Solid
Percent Purity 98%
Chemical Name or Material 2-Fluoro-4-nitrotoluene, 98%
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

EINECSNumber : 215-845-2

TSCA : TSCA