Alfa Aesar™ 2-Fluoro-4-(pentafluorothio)benzyl alcohol, 97%

Manufacturer: Alfa Aesar™ H3428803

Quantity 1g

 View more versions of this product

Catalog No. AAH3428803

  Check Availability
Add to Cart

    Chemical Identifiers

    Molecular Formula C7H6F6OS
    Formula Weight 252.17
    MDL Number MFCD16652506
    Synonym 2-Fluoro-4-(pentafluorosulfur)benzyl alcohol
    Chemical Name or Material 2-Fluoro-4-(pentafluorothio)benzyl alcohol

    Specifications

    Formula Weight 252.17
    Molecular Formula C7H6F6OS
    MDL Number MFCD16652506
    Synonym 2-Fluoro-4-(pentafluorosulfur)benzyl alcohol
    Quantity 1g
    Chemical Name or Material 2-Fluoro-4-(pentafluorothio)benzyl alcohol
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit