Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2'-Fluoro-4'-(trifluoromethyl)acetophenone, 97%

Click to view available options
Quantity:
1g
5g
122023-29-4
C9H6F4O
206.14
MFCD00061247
OLDXVWXLMGJVPH-UHFFFAOYSA-N
2'-fluoro-4'-trifluoromethyl acetophenone,2-fluoro-4-trifluoromethyl acetophenone,1-2-fluoro-4-trifluoromethyl phenyl ethanone,ethanone, 1-2-fluoro-4-trifluoromethyl phenyl,1-acetyl-2-fluoro-4-trifluoromethyl benzene,pubchem8406,acmc-1c256,1-2-fluoro-4-trifluoromethyl-phenyl-ethanone
145568
1-[2-fluoro-4-(trifluoromethyl)phenyl]ethanone
CC(=O)C1=C(C=C(C=C1)C(F)(F)F)F
122023-29-4
C9H6F4O
206.14
MFCD00061247
OLDXVWXLMGJVPH-UHFFFAOYSA-N
2'-fluoro-4'-trifluoromethyl acetophenone,2-fluoro-4-trifluoromethyl acetophenone,1-2-fluoro-4-trifluoromethyl phenyl ethanone,ethanone, 1-2-fluoro-4-trifluoromethyl phenyl,1-acetyl-2-fluoro-4-trifluoromethyl benzene,pubchem8406,acmc-1c256,1-2-fluoro-4-trifluoromethyl-phenyl-ethanone
145568
1-[2-fluoro-4-(trifluoromethyl)phenyl]ethanone
CC(=O)C1=C(C=C(C=C1)C(F)(F)F)F
Quantity 5g
Formula Weight 206.14
Percent Purity 97%
Chemical Name or Material 2'-Fluoro-4'-(trifluoromethyl)acetophenone

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning