Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Fluoro-4-(trifluoromethyl)benzyl bromide, 97%

Click to view available options
Quantity:
1g
5g
239087-07-1
C8H5BrF4
257.026
MFCD00061173
MQUCRCQNRDHRPL-UHFFFAOYSA-N
1-bromomethyl-2-fluoro-4-trifluoromethyl benzene,2-fluoro-4-trifluoromethyl benzyl bromide,2-fluoro-4-trifluoromethylbenzyl bromide,acmc-1ch3e,4-bromomethyl-3-fluorobenzotrifluoride,2-fluoro-4-trifluoromethyl benzylbromide,2-fluoro-4-trifluoromethyl-benzyl bromide,benzene,1-bromomethyl-2-fluoro-4-trifluoromethyl,a-bromo-2-fluoro-4-trifluoromethyl toluene;a-bromo-2-fluoro-4-trifluoromethyl toluene
2737570
1-(bromomethyl)-2-fluoro-4-(trifluoromethyl)benzene
C1=CC(=C(C=C1C(F)(F)F)F)CBr
239087-07-1
C8H5BrF4
257.026
MFCD00061173
MQUCRCQNRDHRPL-UHFFFAOYSA-N
1-bromomethyl-2-fluoro-4-trifluoromethyl benzene,2-fluoro-4-trifluoromethyl benzyl bromide,2-fluoro-4-trifluoromethylbenzyl bromide,acmc-1ch3e,4-bromomethyl-3-fluorobenzotrifluoride,2-fluoro-4-trifluoromethyl benzylbromide,2-fluoro-4-trifluoromethyl-benzyl bromide,benzene,1-bromomethyl-2-fluoro-4-trifluoromethyl,a-bromo-2-fluoro-4-trifluoromethyl toluene;a-bromo-2-fluoro-4-trifluoromethyl toluene
2737570
1-(bromomethyl)-2-fluoro-4-(trifluoromethyl)benzene
C1=CC(=C(C=C1C(F)(F)F)F)CBr
Melting Point 28°C to 30°C
Quantity 1g
UN Number UN1759
Formula Weight 257.03
Percent Purity 97%
Chemical Name or Material 2-Fluoro-4-(trifluoromethyl)benzyl bromide

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.