Alfa Aesar™ 2-Fluoro-4-(trifluoromethyl)pyridine, 97%

Manufacturer: Alfa Aesar™ H33198-03

Quantity 1g

 View more versions of this product

Catalog No. AAH3319803

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 118078-66-3
    Molecular Formula C6H3F4N
    Formula Weight 165.09g/mol
    MDL Number MFCD09879262
    Chemical Name or Material 2-Fluoro-4-(trifluoromethyl)pyridine

    Specifications

    Boiling Point 110°C
    Quantity 1g
    CAS 118078-66-3
    Molecular Formula C6H3F4N
    Formula Weight 165.09g/mol
    MDL Number MFCD09879262
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material 2-Fluoro-4-(trifluoromethyl)pyridine
    Assay 97%

    Description

    Applications
    2-Fluoro-4-(trifluoromethyl)pyridine acts as a reactant in the preparation of aminopyridines through amination reactions. It is also used as a catalytic ligand for regioselective preparation of tetramethylbiphenyls through aerobic oxidative coupling of xylene catalyzed by palladium.

    Solubility
    Miscible with water, alcohol, acetonitrile and N,N-dimethylformamide

    Notes
    Incompatible with oxidizing agents and bases.

    GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit