Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoro-5-formylbenzonitrile 98.0+%, TCI America™
SDP

Catalog No. p-7125597
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 218301-22-5
Molecular Formula C8H4FNO
Molecular Weight (g/mol) 149.124
MDL Number MFCD01863558
InChI Key MOFRJTLODZILCR-UHFFFAOYSA-N
Synonym 3-cyano-4-fluorobenzaldehyde, 4-fluoro-3-cyanobenzaldehyde, 2-fluoro-5-formyl-benzonitrile, benzonitrile, 2-fluoro-5-formyl, 2-fluoro-5-formylbenzenecarbonitrile, 2-fluor-5-formylbenzonitrile, 10014 2-fluoro-5-formyl benzonitrile, pubchem2252, acmc-1cfa9, fluoro-5-formylbenzonitrile
PubChem CID 2769582
IUPAC Name 2-fluoro-5-formylbenzonitrile
SMILES C1=CC(=C(C=C1C=O)C#N)F

Specifications

Melting Point 85°C
Color Yellow
Quantity 1 g
Formula Weight 149.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Fluoro-5-formylbenzonitrile
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.