Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 2-Fluoro-5-methylbenzenesulfonyl chloride, 97%
Click to view available options
Quantity:
1g
5g
Specifications
Melting Point | 32°C to 34°C |
Sensitivity | Moisture sensitive |
Quantity | 5g |
UN Number | UN3261 |
Formula Weight | 208.64 |
Percent Purity | 97% |
Chemical Name or Material | 2-Fluoro-5-methylbenzenesulfonyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures