Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoro-5-methylpyridine, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
2369-19-9
C6H6FN
111.12
AOSOZARHUJMBLZ-UHFFFAOYSA-N
2-fluoro-5-picoline,6-fluoro-3-picoline,2-fluoro-5-methypyridine,2-fluoro-5-methyl-pyridine,fluoromethylpyridine3,2-fluoro-5-methyl pyridine,pyridine, 2-fluoro-5-methyl,pubchem4059,acmc-1cdos,5-methyl-2-fluoropyridine
2774635
2-fluoro-5-methylpyridine
CC1=CN=C(C=C1)F
2369-19-9
C6H6FN
111.12
AOSOZARHUJMBLZ-UHFFFAOYSA-N
2-fluoro-5-picoline,6-fluoro-3-picoline,2-fluoro-5-methypyridine,2-fluoro-5-methyl-pyridine,fluoromethylpyridine3,2-fluoro-5-methyl pyridine,pyridine, 2-fluoro-5-methyl,pubchem4059,acmc-1cdos,5-methyl-2-fluoropyridine
2774635
2-fluoro-5-methylpyridine
CC1=CN=C(C=C1)F
Packaging Glass Bottle
Flash Point 48°C
Color Colorless to Yellow
Quantity 1g
Boiling Point 154.0°C to 155.0°C
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Formula Weight 111.12
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 2-Fluoro-5-methylpyridine
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapour.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning