Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoro-5-nitrotoluene, 97%, ACROS Organics™

Click to view available options
Quantity:
10g
Packaging:
Glass bottle
455-88-9
C7H6FNO2
155.128
MFCD00007284
XUCYJGMIICONES-UHFFFAOYSA-N
2-fluoro-5-nitrotoluene,benzene, 1-fluoro-2-methyl-4-nitro,4-fluoro-3-methylnitrobenzene,toluene, 2-fluoro-5-nitro,1-fluoro-2-methyl-4-nitro-benzene,5-nitro-2-fluorotoluene,3-methyl-4-fluoronitrobenzene,pubchem1593,2-fluoro-5-nitro-toluene,5-nitro-2-fluoro toluene
68001
1-fluoro-2-methyl-4-nitrobenzene
CC1=C(C=CC(=C1)[N+](=O)[O-])F
455-88-9
C7H6FNO2
155.128
MFCD00007284
XUCYJGMIICONES-UHFFFAOYSA-N
2-fluoro-5-nitrotoluene,benzene, 1-fluoro-2-methyl-4-nitro,4-fluoro-3-methylnitrobenzene,toluene, 2-fluoro-5-nitro,1-fluoro-2-methyl-4-nitro-benzene,5-nitro-2-fluorotoluene,3-methyl-4-fluoronitrobenzene,pubchem1593,2-fluoro-5-nitro-toluene,5-nitro-2-fluoro toluene
68001
1-fluoro-2-methyl-4-nitrobenzene
CC1=C(C=CC(=C1)[N+](=O)[O-])F
Quantity 10g
Color Yellow
Infrared Spectrum Authentic
Melting Point 36°C to 41°C
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula CH3C6H3(NO2)F
Beilstein 05, II, 251
Formula Weight 155.13
Physical Form Crystals or Crystalline Chunks
Percent Purity ≥96.0%
Chemical Name or Material 2-Fluoro-5-nitrotoluene
Show More Show Less

GHS P Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
Harmful in contact with skin.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning