Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Fluoro-5-nitrotoluene, 98+%

Click to view available options
Quantity:
10g
50g
455-88-9
C7H6FNO2
155.13
MFCD00007284
XUCYJGMIICONES-UHFFFAOYSA-N
2-fluoro-5-nitrotoluene, benzene, 1-fluoro-2-methyl-4-nitro, 4-fluoro-3-methylnitrobenzene, toluene, 2-fluoro-5-nitro, 1-fluoro-2-methyl-4-nitro-benzene, 5-nitro-2-fluorotoluene, 3-methyl-4-fluoronitrobenzene, pubchem1593, 2-fluoro-5-nitro-toluene, 5-nitro-2-fluoro toluene
68001
1-fluoro-2-methyl-4-nitrobenzene
CC1=CC(=CC=C1F)[N+]([O-])=O
455-88-9
C7H6FNO2
155.13
MFCD00007284
XUCYJGMIICONES-UHFFFAOYSA-N
2-fluoro-5-nitrotoluene, benzene, 1-fluoro-2-methyl-4-nitro, 4-fluoro-3-methylnitrobenzene, toluene, 2-fluoro-5-nitro, 1-fluoro-2-methyl-4-nitro-benzene, 5-nitro-2-fluorotoluene, 3-methyl-4-fluoronitrobenzene, pubchem1593, 2-fluoro-5-nitro-toluene, 5-nitro-2-fluoro toluene
68001
1-fluoro-2-methyl-4-nitrobenzene
CC1=CC(=CC=C1F)[N+]([O-])=O
Quantity 10g
Flash Point 105°C (221°F)
Melting Point 37°C to 40°C
Boiling Point 98°C to 101°C (13mmHg)
UN Number UN2811
Beilstein 1940341
Formula Weight 155.13
Percent Purity ≥98%
Chemical Name or Material 2-Fluoro-5-nitrotoluene

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 207-251-7

TSCA : Yes

Recommended Storage : Ambient temperatures