Alfa Aesar™ 2-Fluoro-5-(trifluoromethoxy)aniline, 97%

Manufacturer: Alfa Aesar™ H3210306

Quantity 5g

 View more versions of this product

Catalog No. AAH3210306

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 116369-23-4
    Molecular Formula C7H5F4NO
    Formula Weight 195.12
    MDL Number MFCD09832254
    UN Number UN2810
    Chemical Name or Material 2-Fluoro-5-(trifluoromethoxy)aniline

    Specifications

    Formula Weight 195.12
    Molecular Formula C7H5F4NO
    MDL Number MFCD09832254
    CAS 116369-23-4
    UN Number UN2810
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H311-H302-H332-H315-H319
    Toxic in contact with skin.
    Harmful if swallowed.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338-P309-P310-P302+P352
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN: Wash with plenty of soap and water.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
    Quantity 5g
    Chemical Name or Material 2-Fluoro-5-(trifluoromethoxy)aniline
    Assay 97%

    Description

    Applications
    In discovery of novel benzimidazoles as potent inhibitors of TIE-2 and VEGFR-2 tyrosine kinase receptors 2-Fluoro-5-(trifluoromethoxy)aniline is used as coupling agent. 2-Fluoro-5-(trifluoromethyl)aniline may be used in chemical synthesis studies.

    Solubility
    Insoluble in water.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

    GHS H Statement
    H311-H302-H332-H315-H319
    Toxic in contact with skin.
    Harmful if swallowed.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280h-P305+P351+P338-P309-P310-P302+P352
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN: Wash with plenty of soap and water.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit