Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Fluoro-5-(trifluoromethyl)benzaldehyde, 97%

Click to view available options
Quantity:
1g
5g
25g
146137-78-2
C8H4F4O
192.113
MFCD00061311
IDLNLGMUINCSGS-UHFFFAOYSA-N
2-fluoro-5-trifluoromethyl benzaldehyde,2-fluoro-5-trifluoromethyl-benzaldehyde,4-fluoro-3-formylbenzotrifluoride,benzaldehyde, 2-fluoro-5-trifluoromethyl,pubchem5798,acmc-209cwf,intermediates-zcf02118,ksc496a6r
2734904
2-fluoro-5-(trifluoromethyl)benzaldehyde
C1=CC(=C(C=C1C(F)(F)F)C=O)F
146137-78-2
C8H4F4O
192.113
MFCD00061311
IDLNLGMUINCSGS-UHFFFAOYSA-N
2-fluoro-5-trifluoromethyl benzaldehyde,2-fluoro-5-trifluoromethyl-benzaldehyde,4-fluoro-3-formylbenzotrifluoride,benzaldehyde, 2-fluoro-5-trifluoromethyl,pubchem5798,acmc-209cwf,intermediates-zcf02118,ksc496a6r
2734904
2-fluoro-5-(trifluoromethyl)benzaldehyde
C1=CC(=C(C=C1C(F)(F)F)C=O)F
Sensitivity Air Sensitive
Quantity 1g
Density 1.36
Refractive Index 1.451
Boiling Point 176°C
Flash Point 76°C (168°F)
Beilstein 5810686
Formula Weight 192.11
Percent Purity 97%
Chemical Name or Material 2-Fluoro-5-(trifluoromethyl)benzaldehyde

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures