Alfa Aesar™ 2-Fluoro-5-(trifluoromethyl)benzenesulfonyl chloride, 97%

Manufacturer: Alfa Aesar™ H26687MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH26687MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1744-43-0
    Molecular Formula C7H3ClF4O2S
    Formula Weight 262.61
    MDL Number MFCD01090999
    UN Number UN3265
    Chemical Name or Material 2-Fluoro-5-(trifluoromethyl)benzenesulfonyl chloride

    Specifications

    Formula Weight 262.61
    Molecular Formula C7H3ClF4O2S
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
    Quantity 250mg
    MDL Number MFCD01090999
    CAS 1744-43-0
    UN Number UN3265
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material 2-Fluoro-5-(trifluoromethyl)benzenesulfonyl chloride
    Assay 97%

    Description

    Applications
    2-Fluoro-5-(trifluoromethyl)benzenesulfonyl chloride is used to describe a straight or branched chain alkyl.

    Solubility
    Slightly soluble in water.

    Notes
    Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit