Alfa Aesar™ 2-Fluoro-5-(trifluoromethyl)pyridine, 97%

Manufacturer: Alfa Aesar™ H3257403

Quantity 1g

 View more versions of this product

Catalog No. AAH3257403

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 69045-82-5
    Molecular Formula C6H3F4N
    Formula Weight 165.09
    MDL Number MFCD07437945
    UN Number UN1993
    Synonym alpha,alpha,alpha,6-Tetrafluoro-3-picoline
    Chemical Name or Material 2-Fluoro-5-(trifluoromethyl)pyridine

    Specifications

    Formula Weight 165.09
    Quantity 1g
    Molecular Formula C6H3F4N
    MDL Number MFCD07437945
    CAS 69045-82-5
    UN Number UN1993
    Synonym alpha,alpha,alpha,6-Tetrafluoro-3-picoline
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H226-H317
    Flammable liquid and vapor.
    May cause an allergic skin reaction.
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    Flash Point 35°C (95°F)
    Boiling Point 118° to 122°C
    Chemical Name or Material 2-Fluoro-5-(trifluoromethyl)pyridine
    Assay 97%

    GHS H Statement
    H226-H317
    Flammable liquid and vapor.
    May cause an allergic skin reaction.

    GHS P Statement
    P210-P261-P280-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit