Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoro-6-methoxybenzaldehyde, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
146137-74-8
C8H7FO2
154.14
UIOAYOIJMYMOEU-UHFFFAOYSA-N
2-methoxy-6-fluorobenzaldehyde,6-fluoro-o-anisaldehyde,2-fluoro-6-methoxy-benzaldehyde,2-fluoro-6-mehoxybenzaldehyde,benzaldehyde, 2-fluoro-6-methoxy,2-fluoro-6-methoxybenzaldehyde,pubchem8486,acmc-209yud,3-fluoro-2-formylanisole,intermediates-zcf02117
2737357
2-fluoro-6-methoxybenzaldehyde
COC1=C(C(=CC=C1)F)C=O
146137-74-8
C8H7FO2
154.14
UIOAYOIJMYMOEU-UHFFFAOYSA-N
2-methoxy-6-fluorobenzaldehyde,6-fluoro-o-anisaldehyde,2-fluoro-6-methoxy-benzaldehyde,2-fluoro-6-mehoxybenzaldehyde,benzaldehyde, 2-fluoro-6-methoxy,2-fluoro-6-methoxybenzaldehyde,pubchem8486,acmc-209yud,3-fluoro-2-formylanisole,intermediates-zcf02117
2737357
2-fluoro-6-methoxybenzaldehyde
COC1=C(C(=CC=C1)F)C=O
Quantity 5g
Infrared Spectrum Authentic
Flash Point 110°C
Color Tan to White
Melting Point 57.0°C to 60.0°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Formula Weight 154.14
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 2-Fluoro-6-methoxybenzaldehyde
Show More Show Less

GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning