Alfa Aesar™ 2'-Fluoro-6'-(trifluoromethyl)acetophenone, 98%

Manufacturer: Alfa Aesar™ B2450106

Quantity 5g

 View more versions of this product

Catalog No. AAB2450106

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 174013-29-7
    Molecular Formula C9H6F4O
    Formula Weight 206.14
    MDL Number MFCD00061249
    Beilstein 9193675
    Chemical Name or Material 2'-Fluoro-6'-(trifluoromethyl)acetophenone

    Specifications

    Formula Weight 206.14
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.
    Quantity 5g
    Molecular Formula C9H6F4O
    MDL Number MFCD00061249
    CAS 174013-29-7
    Beilstein 9193675
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Flash Point 86°C (186°F)
    Chemical Name or Material 2'-Fluoro-6'-(trifluoromethyl)acetophenone
    Assay 98%

    GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.

    GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit