Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-Fluoro-6-(trifluoromethyl)benzaldehyde, 98%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Specifications
Boiling Point | 156°C |
Flash Point | 82°C |
Assay Percent Range | 97.5% min. (GC) |
Packaging | Glass Bottle |
Quantity | 1 g |
Formula Weight | 192.11 |
Percent Purity | 98% |
Chemical Name or Material | 2-Fluoro-6-(trifluoromethyl)benzaldehyde |
Safety and Handling
Hazard Category | GHS Signal Word: Warning |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction