Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Fluoro-6-(trifluoromethyl)benzoic acid, 98%

Click to view available options
Quantity:
1g
5g
32890-94-1
C8H4F4O2
208.112
MFCD00040981
LNARMXLVVGHCRP-UHFFFAOYSA-N
2-fluoro-6-trifluoromethyl benzoic acid,otf-boa-6f,alpha,alpha,alpha,6-tetrafluoro-o-toluic acid,benzoic acid, 2-fluoro-6-trifluoromethyl,6-fluoro-2-trifluoromethylbenzoic acid,2-fluoro-6-trifluoromethyl-benzoic acid,6-fluoro-2-trifluoromethyl benzoic acid,pubchem4759,acmc-20a40d,rarechem al bo 0626
688123
2-fluoro-6-(trifluoromethyl)benzoic acid
C1=CC(=C(C(=C1)F)C(=O)O)C(F)(F)F
32890-94-1
C8H4F4O2
208.112
MFCD00040981
LNARMXLVVGHCRP-UHFFFAOYSA-N
2-fluoro-6-trifluoromethyl benzoic acid,otf-boa-6f,alpha,alpha,alpha,6-tetrafluoro-o-toluic acid,benzoic acid, 2-fluoro-6-trifluoromethyl,6-fluoro-2-trifluoromethylbenzoic acid,2-fluoro-6-trifluoromethyl-benzoic acid,6-fluoro-2-trifluoromethyl benzoic acid,pubchem4759,acmc-20a40d,rarechem al bo 0626
688123
2-fluoro-6-(trifluoromethyl)benzoic acid
C1=CC(=C(C(=C1)F)C(=O)O)C(F)(F)F
Quantity 5g
Melting Point 83°C to 88°C
Beilstein 2112882
Formula Weight 208.11
Percent Purity 98%
Chemical Name or Material 2-Fluoro-6-(trifluoromethyl)benzoic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning