Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoro-6-(trifluoromethyl)benzyl bromide, 95%, Acros Organics

Click to view available options
Quantity:
1g
5g
239087-08-2
C8H5BrF4
257.03
RINUERVPFANASB-UHFFFAOYSA-N
2-fluoro-6-trifluoromethyl benzyl bromide,2-bromomethyl-1-fluoro-3-trifluoromethyl benzene,2-fluoro-6-trifluoromethylbenzyl bromide,2-fluoro-6-trifluoromethylbenzylbromide,2-fluoro-6-trifluoromethyl benzylbromide,2-bromomethyl-3-fluorobenzotrifluoride,alpha-bromo-2-fluoro-6-trifluoromethyl toluene,benzene, 2-bromomethyl-1-fluoro-3-trifluoromethyl,pubchem4932,acmc-1cg83
2737573
2-(bromomethyl)-1-fluoro-3-(trifluoromethyl)benzene
C1=CC(=C(C(=C1)F)CBr)C(F)(F)F
239087-08-2
C8H5BrF4
257.03
RINUERVPFANASB-UHFFFAOYSA-N
2-fluoro-6-trifluoromethyl benzyl bromide,2-bromomethyl-1-fluoro-3-trifluoromethyl benzene,2-fluoro-6-trifluoromethylbenzyl bromide,2-fluoro-6-trifluoromethylbenzylbromide,2-fluoro-6-trifluoromethyl benzylbromide,2-bromomethyl-3-fluorobenzotrifluoride,alpha-bromo-2-fluoro-6-trifluoromethyl toluene,benzene, 2-bromomethyl-1-fluoro-3-trifluoromethyl,pubchem4932,acmc-1cg83
2737573
2-(bromomethyl)-1-fluoro-3-(trifluoromethyl)benzene
C1=CC(=C(C(=C1)F)CBr)C(F)(F)F
Melting Point 42°C to 44°C
Flash Point 107°C
Quantity 5g
Assay Percent Range 94% min. (GC)
Formula Weight 257.03
Percent Purity 95%
Chemical Name or Material 2-Fluoro-6-(trifluoromethyl)benzyl bromide

GHS Signal Word: Danger