Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Fluoro-6-(trifluoromethyl)pyridine, 98%

Manufacturer:  Alfa Aesar™ H3263303

Quantity 1g

 View more versions of this product

Catalog No. AAH3263303


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

94239-04-0
165.09
UN1993
C6H3F4N
MFCD03092901
2-Fluoro-6-(trifluoromethyl)pyridine

Specifications

1g
C6H3F4N
MFCD03092901
2-Fluoro-6-(trifluoromethyl)pyridine
94239-04-0
165.09
UN1993
98%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H226-H302-H332
Flammable liquid and vapor.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement
P210-P261-P280-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit