Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-Fluoro-9H-carbazole, TCI America™
Click to view available options
Quantity:
200mg
1g
Specifications
Quantity | 200mg |
Color | Undesignated |
Melting Point | 227°C |
Formula Weight | 185.20 |
Physical Form | Crystal-Powder at 20°C |
Percent Purity | ≥98.0% (GC) |
Chemical Name or Material | 2-Fluoro-9H-carbazole |
Safety and Handling
TSCA : No