Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoro-alpha-methyl-4-biphenylacetic acid, 99%, Thermo Scientific Chemicals

Catalog Number AAB2260303
Click to view available options
Quantity:
1 g
5 g
25 g
5104-49-4
C15H13FO2
244.27
MFCD00079303
SYTBZMRGLBWNTM-UHFFFAOYNA-N
flurbiprofen, ansaid, froben, antadys, cebutid, flurofen, anside, flurbiprofene, flurbiprofeno, flurbiprofenum
3394
CHEBI:5130
CC(C(O)=O)C1=CC=C(C(F)=C1)C1=CC=CC=C1
5104-49-4
C15H13FO2
244.27
MFCD00079303
SYTBZMRGLBWNTM-UHFFFAOYNA-N
flurbiprofen, ansaid, froben, antadys, cebutid, flurofen, anside, flurbiprofene, flurbiprofeno, flurbiprofenum
3394
CHEBI:5130
CC(C(O)=O)C1=CC=C(C(F)=C1)C1=CC=CC=C1

A cyclooxygenase inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 112°C to 114°C
Odor Odorless
Quantity 1 g
UN Number UN2811
Merck Index 14,4199
IUPAC Name 2-(3-fluoro-4-phenylphenyl)propanoic acid
Formula Weight 244.27
Percent Purity 99%
Chemical Name or Material 2-Fluoro-alpha-methyl-4-biphenylacetic acid
Hazard Category H301-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 225-827-6
RTECSNumber DU8341000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit