Alfa Aesar™ 2-Fluoro-D-phenylalanine, 99+% (sum of enantiomers)

Manufacturer: Alfa Aesar™ H2752403

Quantity 1g

 View more versions of this product

Catalog No. AAH2752403

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 97731-02-7
    Molecular Formula C9H10FNO2
    Formula Weight 183.18
    MDL Number MFCD00066447
    Synonym H-D-Phe(2-F)-OH; o-Fluoro-D-phenylalanine
    Chemical Name or Material 2-Fluoro-D-phenylalanine, sum of enantiomers

    Specifications

    Formula Weight 183.18
    MDL Number MFCD00066447
    CAS 97731-02-7
    Synonym H-D-Phe(2-F)-OH; o-Fluoro-D-phenylalanine
    Melting Point 209° to 211°C
    Quantity 1g
    Molecular Formula C9H10FNO2
    Chemical Name or Material 2-Fluoro-D-phenylalanine, sum of enantiomers
    Assay ≥99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit