Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Fluoro-m-xylene, 98%

Manufacturer:  Alfa Aesar™ A1042406

Quantity 5g

 View more versions of this product

Catalog No. AAA1042406


  Check Availability
Add to cart

Specifications

Description

Applications
It is used in the spectroscopic observation of jet-cooled 2-fluoro-m-xylyl radical.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from electrostatic charges and keep away from ignition sources.
Description & Specifications

Chemical Identifiers

443-88-9
124.16
UN1993
2,6-Dimethylfluorobenzene; 2-Fluoro-1.3-dimethylbenzene
C8H9F
MFCD00039215
1857413
2-Fluoro-m-xylene

Specifications

30°C (86°F)
147° to 148°C
443-88-9
124.16
UN1993
2,6-Dimethylfluorobenzene; 2-Fluoro-1.3-dimethylbenzene
98%
Aromatic
5g
C8H9F
MFCD00039215
1857413
2-Fluoro-m-xylene

Safety and Handling

SDS & Safety Documents

GHS H Statement
H226
Flammable liquid and vapor.

GHS P Statement
P261
Avoid breathing dust/fume/gas/mist/vapors/spray.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit