Alfa Aesar™ 2-Fluoro-N-methylbenzylamine, 95%

Manufacturer: Alfa Aesar™ H5443903

Quantity 1g

 View more versions of this product

Catalog No. AAH5443903

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 399-30-4
    Formula Weight 139.17
    MDL Number MFCD04629633
    Chemical Name or Material 2-Fluoro-N-methylbenzylamine

    Specifications

    Formula Weight 139.17
    Health Hazard 3 GHS P Statement
    P210-P280-P305+P351+P338-P362-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 1 Warning
    Boiling Point 73°C
    Physical Form Liquid
    MDL Number MFCD04629633
    Refractive Index 1.047
    Density 1.047g/mL
    Quantity 1g
    CAS 399-30-4
    Health Hazard 2 GHS H Statement
    H315-H319-H227
    Causes skin irritation.
    Causes serious eye irritation.
    Combustible liquid.
    Sensitivity Air Sensitive
    Color Colorless to tan
    Chemical Name or Material 2-Fluoro-N-methylbenzylamine
    Assay 95%

    Description

    Applications
    It is used as a pharmaceutical intermediate.

    Solubility
    Slightly soluble in water.

    Notes
    Incompatible with strong oxidizing agents and air. Store at 4-2°C.

    GHS H Statement
    H315-H319-H227
    Causes skin irritation.
    Causes serious eye irritation.
    Combustible liquid.

    GHS P Statement
    P210-P280-P305+P351+P338-P362-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit