Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2'-Fluoroacetanilide, 98+%

Click to view available options
Quantity:
25g
100g
399-31-5
C8H8FNO
153.156
MFCD00000315
AUZPZBPZWHEIDY-UHFFFAOYSA-N
2'-fluoroacetanilide,n-2-fluorophenyl acetamide,acetamide, n-2-fluorophenyl,a-fluoroacetanilide,pubchem3369,maybridge1_008728,n-acetyl-2-fluoroaniline,acmc-1aho3,n-2-fluorophenyl acetamide #
67860
N-(2-fluorophenyl)acetamide
CC(=O)NC1=CC=CC=C1F
399-31-5
C8H8FNO
153.156
MFCD00000315
AUZPZBPZWHEIDY-UHFFFAOYSA-N
2'-fluoroacetanilide,n-2-fluorophenyl acetamide,acetamide, n-2-fluorophenyl,a-fluoroacetanilide,pubchem3369,maybridge1_008728,n-acetyl-2-fluoroaniline,acmc-1aho3,n-2-fluorophenyl acetamide #
67860
N-(2-fluorophenyl)acetamide
CC(=O)NC1=CC=CC=C1F
Boiling Point 140°C to 142°C (14mmHg)
Quantity 100g
Melting Point 76°C to 80°C
Beilstein 2717443
Formula Weight 153.16
Percent Purity ≥98%
Chemical Name or Material 2'-Fluoroacetanilide

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 206-916-9

TSCA : No

Recommended Storage : Ambient temperatures