Alfa Aesar™ 2-Fluoroanisole, 98%

Manufacturer: Alfa Aesar™ A12380-14

Quantity 25g

 View more versions of this product

Catalog No. AAA1238014

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 321-28-8
    Molecular Formula C7H7FO
    Formula Weight 126.13g/mol
    MDL Number MFCD00000316
    UN Number UN1993
    Beilstein 1859755
    Chemical Name or Material 2-Fluoroanisole

    Specifications

    Melting Point −39°C
    Boiling Point 154° to 155°C
    Quantity 25g
    Flash Point 56°C (133°F)
    CAS 321-28-8
    Molecular Formula C7H7FO
    Formula Weight 126.13g/mol
    MDL Number MFCD00000316
    UN Number UN1993
    Beilstein 1859755
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H226
    Flammable liquid and vapor.
    Health Hazard 3 GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    Chemical Name or Material 2-Fluoroanisole
    Assay 98%

    GHS H Statement
    H226
    Flammable liquid and vapor.

    GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit